Learn More
Cayman Chemical MST-AS5568 L-681 217 2.5mg
SDP
Catalog No. 502936235
Encompass_Preferred
An antibiotic; active against Gram positive and Gram negative bacteria at 255 UG/ml; inhibits bacterial protein synthesis (EC50 = 0.4 µM in E. coli); increases EF-Tu-associated GTPase activity (EC50 = 6 µM in E. coli)
Catalog No. 50-293-6235 Supplier Cayman Chemical Supplier No. 266892.5 MG
An antibiotic; active against Gram positive and Gram negative bacteria at 255 UG/ml; inhibits bacterial protein synthesis (EC50 = 0.4 µM in E. coli); increases EF-Tu-associated GTPase activity (EC50 = 6 µM in E. coli)