Learn More
Aviva Systems Biology Corp MST1 Antibody - middle region (ARP45724_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45724_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MST1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MST1. Alternate names: D3F15S2, DNF15S2, HGFL, MSP, NF15S2ApplicationWB, IHC. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-59119