Learn More
Regis Technologies Inc GC Regeant, Regis, MSTFA (10 x 1 GM)
SDP
Catalog No. 50442916
Encompass
MSTFA is a Silylation Reagent. MSTFA is the most volatile of the TMS-acetamides and useful in the analysis of volatile trace materials. MSTFA is more volatile than BSA or BSTFA but has similar silylation strength.
Catalog No. 50-442-916 Supplier Regis Technologies Inc Supplier No. 1270590200
MSTFA is a Silylation Reagent. MSTFA is the most volatile of the TMS-acetamides and useful in the analysis of volatile trace materials. MSTFA is more volatile than BSA or BSTFA but has similar silylation strength.