Learn More
AVANTOR MASTERFLEX (VWR LLC) Ismatec Pump Tubing, PharMed BPT, 0.19 mm ID; 100 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9580910/DEL
Encompass
The overall length of PharMed BPT 95809-series extension tubing is 100 feet (30 m). The legacy Ismatec extension tubing was 3 m long.
Catalog No. NC1002218