Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, Tygon E-LFL, L/S 25; 25 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0644025/DEL
Encompass
Applications: General laboratory use Characteristics: Long life, won¿t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, transparent
Catalog No. NC0548995