Learn More
OriGene OriGene Human shRNA Lentiviral Plasmid Kit, Targets Myostatin, 70% Knockdown Efficiency Guaranteed, 5 µg
SDP
Catalog No. NC3654239
Encompass_Preferred
The OriGene Technologies Human shRNA Lentiviral Plasmid Kit efficiently targets Myostatin gene expression in human cells, enabling both transient and stable gene knockdown. With a guarantee of ≥70% knockdown efficiency, this kit includes sequence-verified shRNA constructs and a third-generation lentiviral vector, featuring a selection marker and fluorescent reporter for easy identification of transduced cells. It's suitable for researchers looking to effectively manipulate gene expression.
Catalog No. NC3654239 Supplier OriGene Supplier No. TL312799V
The OriGene Technologies Human shRNA Lentiviral Plasmid Kit efficiently targets Myostatin gene expression in human cells, enabling both transient and stable gene knockdown. With a guarantee of ≥70% knockdown efficiency, this kit includes sequence-verified shRNA constructs and a third-generation lentiviral vector, featuring a selection marker and fluorescent reporter for easy identification of transduced cells. It's suitable for researchers looking to effectively manipulate gene expression.