Learn More
Aviva Systems Biology Corp MSTO1 Antibody - N-terminal region (ARP40895_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40895_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MSTO1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MSTO1. Alternate names: DKFZp686B1757, DKFZp686I01261, FLJ10504, LST005, MST, RP11-29H23.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-902