Learn More
ATCC MSTO-211H; Mesothelioma; Human (Homo sapiens)
Catalog No. 502382553
Encompass
MSTO-211H is a cell line exhibiting fibroblast morphology that was isolated in 1985 from the lung of a 62-year-old, White male patient with biphasic mesothelioma. This cell line was deposited by G Bepler.
Catalog No. 50-238-2553 Supplier ATCC Supplier No. CRL2081
MSTO-211H is a cell line exhibiting fibroblast morphology that was isolated in 1985 from the lung of a 62-year-old, White male patient with biphasic mesothelioma. This cell line was deposited by G Bepler.