Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Digital Miniflex Pump, Dual-Channel, 0.1 to 300 rpm; 90 to 260 VAC
Catalog No. NC1132317
Encompass
This compact, stackable pump features a durable ABS housing that is easy to wipe down and keep clean. The Miniflex pump head features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life. Note:Dual-channel model must have same size tubing loaded in both channels to ensure tubing retention.
Catalog No. NC1132317 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0752540
This compact, stackable pump features a durable ABS housing that is easy to wipe down and keep clean. The Miniflex pump head features a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life. Note:Dual-channel model must have same size tubing loaded in both channels to ensure tubing retention.