Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Portable Sampling Pump; 115/230 VAC
Catalog No. NC1831567
Encompass
Masterflex L/S Portable Sampling Pump; 115/230 VAC,Features a single-turn potentiometer for speed control, ON indicator, and three-position power/reversing switchReversible flow and prime/purge functionIncludes a Masterflex L/S Easy-Load pump headAlso accepts Masterflex L/S Standard pump heads (order separately)Floats up to 30 minutes if dropped in water when case is closed and latched
Catalog No. NC1831567 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0757102
Masterflex L/S Portable Sampling Pump; 115/230 VAC,Features a single-turn potentiometer for speed control, ON indicator, and three-position power/reversing switchReversible flow and prime/purge functionIncludes a Masterflex L/S Easy-Load pump headAlso accepts Masterflex L/S Standard pump heads (order separately)Floats up to 30 minutes if dropped in water when case is closed and latched