Masterflex L/S spooled precision pump tubing is a platinum-cured silicone tubing formulated for reproducible flow and long service life in peristaltic pumping applications across pharmaceutical, biotechnology, food, and laboratory settings. Precision extrusion and formulation deliver low compression set, broad chemical compatibility, and options for sterilization.
- Platinum-cured silicone material.
- L/S size 16, inner diameter 3.10 mm (0.122 in).
- Spooled length 500 ft (152.4 m) per pack.
- Maximum pressure 2.7 bar (40 psi).
- Typical flow rate 480 mL/min (L/S 16, depends on pump speed).
- Static temperature range -50°C to 230°C (-58°F to 446°F).
- Dynamic temperature range -40°C to 100°C (-40°F to 212°F).
- Compatible with autoclave, ethylene oxide, and gamma sterilization.
- Intended for pharmaceutical, biotech, food, and laboratory peristaltic pumping applications.