Learn More
Accuflow Systems MSTRFLX TBNG PURIFLEX #14 25FT
Catalog No. 501109143
Encompass_Preferred
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Tubing Puri-Flex, L/S 14, 25 feet Long life tubing for any Masterflex pump used in your pharma and bioprocess applications Applications: Cell and tissue culture work, pharmaceutical biotech
Catalog No. 50-110-9143 Supplier Accuflow Systems Supplier No. 9641914
Manufactured by Cole Parmer and sold by Accuflow. Masterflex Tubing Puri-Flex, L/S 14, 25 feet Long life tubing for any Masterflex pump used in your pharma and bioprocess applications Applications: Cell and tissue culture work, pharmaceutical biotech