Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S High-Performance Precision Pump Tubing, Tygon E-LFL , L/S 15; 25 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0644015/DEL
Encompass
Applications: General laboratory use Characteristics: Long life, won¿t age or oxidize, fits tightly to glass and metal, bends to small radius curves, nontoxic, transparent
Catalog No. NC0457340
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.