Learn More
AVANTOR MASTERFLEX (VWR LLC) Tygon Chemical Tubing, L/S 17, 50 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0647517/DEL
Encompass
Masterflex L/S Tygon Chemical Tubing, L/S 17, 50 ft, Compatible with aggressive chemicalsOutstanding chemical resistanceCost-effective alternative to fluoropolymers or fluoroelastomersFDA compliantNOTE: Tygon Chemical 2001 pump tubing is recommended for use in Easy-Load and Easy-Load II pump heads only.
Catalog No. NC9030650
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.