Learn More
INTACT GENOMICS INC MSU440 Agrobacterium ElectroCompetent Cells 18x50µl
Supplier:  INTACT GENOMICS INC 127736
Encompass
Intact Genomics (ig) MSU440 Agrobacterium ElectroCompetent Cells are made from a specific strain of Rhizobium rhizogenes (formerly Agrobacterium rhizogenes), Agrobacterium rhizogenes (str R) MSU440 Ri (agropine type). Agrobacterium rhizogenes is a soil-borne gram-negative bacterium that can infect most dicotyledons, a few monocotyledons and some gymnosperms. MSU440 Electrocompetent Agrobacterium are optimized for the highest transformation efficiencies and are useful for transgenic operations of corn, tobacco, wormwood, tea tree and other plants. MSU440 Agrobacterium rhizogenes strain contains agrobacterium-type Ri plasmid and displays streptomycin resistance.Product Includes and Storage:MSU440 Agrobacterium ElectroCompetent Cells: -80 ºCpCAMBIA1391z control DNA: -20 ºCAgro Recovery medium: 4 ºC
Catalog No. 50-000-83058