Learn More
Medchemexpress LLC MSX-122 | 897657-95-3 | 98.2% | 292.34 | 1 ML
SDP
Catalog No. 5000350285
Encompass_Preferred
MSX-122 is an orally active partial antagonist of CXCR4, inhibiting CXCR4/CXCL12 actions with an IC50 of approximately 10 nM. This compound demonstrates anti-inflammatory and anti-metastatic activity. It is intended for research use only.
  • Orally active partial antagonist of CXCR4
  • Inhibits CXCR4/CXCL12 actions
  • Exhibits anti-inflammatory activity
  • Exhibits anti-metastatic activity
Catalog No. 50-003-50285 Supplier Medchemexpress LLC Supplier No. HY1369610MM/1ML
MSX-122 is an orally active partial antagonist of CXCR4, inhibiting CXCR4/CXCL12 actions with an IC50 of approximately 10 nM. This compound demonstrates anti-inflammatory and anti-metastatic activity. It is intended for research use only.
  • Orally active partial antagonist of CXCR4
  • Inhibits CXCR4/CXCL12 actions
  • Exhibits anti-inflammatory activity
  • Exhibits anti-metastatic activity