Learn More
Medchemexpress LLC Msx-122 | 897657-95-3 | 98.2% | 292.34 | 100 MG
SDP
Catalog No. 5000349010
Encompass_Preferred
MSX-122 is an orally active partial antagonist of CXCR4, effectively inhibiting CXCR4/CXCL12 actions with an IC50 of approximately 10 nM. This compound exhibits both anti-inflammatory and anti-metastatic activities. It potently blocks the invasion of MDA-MB-231 cells in vitro and has been shown to reduce inflammation and lung fibrosis in mice, as well as blocking metastasis in experimental animal models of breast cancer.
  • Orally active partial antagonist of CXCR4.
  • Inhibits CXCR4/CXCL12 actions.
  • Shows anti-inflammatory activity.
  • Exhibits anti-metastatic activity.
  • Potently blocks invasion of certain cancer cells.
  • Decreases hepatic micrometastases in animal models.
Catalog No. 50-003-49010 Supplier Medchemexpress LLC Supplier No. HY13696100MG
MSX-122 is an orally active partial antagonist of CXCR4, effectively inhibiting CXCR4/CXCL12 actions with an IC50 of approximately 10 nM. This compound exhibits both anti-inflammatory and anti-metastatic activities. It potently blocks the invasion of MDA-MB-231 cells in vitro and has been shown to reduce inflammation and lung fibrosis in mice, as well as blocking metastasis in experimental animal models of breast cancer.
  • Orally active partial antagonist of CXCR4.
  • Inhibits CXCR4/CXCL12 actions.
  • Shows anti-inflammatory activity.
  • Exhibits anti-metastatic activity.
  • Potently blocks invasion of certain cancer cells.
  • Decreases hepatic micrometastases in animal models.