Learn More
Medchemexpress LLC MSX-122 | 897657-95-3 | 98.2% | 292.34 | 25 MG
SDP
Catalog No. 5000350624
Encompass_Preferred
MSX-122 is an orally active partial antagonist of CXCR4, inhibiting CXCR4/CXCL12 actions, with an IC50 of ~10 nM.
  • Anti-inflammatory activity.
  • Anti-metastatic activity.
  • Orally active.
  • Potently blocks invasion of MDA-MB-231 cells in vitro.
  • Blocks inflammation induced by carrageenan and lung fibrosis induced by bleomycin in mice.
  • Blocks metastasis in an experimental animal model of breast cancer metastasis.
  • Significantly decreases the numbers of hepatic micrometastases in vivo.
Catalog No. 50-003-50624 Supplier Medchemexpress LLC Supplier No. 25MGHY13696
MSX-122 is an orally active partial antagonist of CXCR4, inhibiting CXCR4/CXCL12 actions, with an IC50 of ~10 nM.
  • Anti-inflammatory activity.
  • Anti-metastatic activity.
  • Orally active.
  • Potently blocks invasion of MDA-MB-231 cells in vitro.
  • Blocks inflammation induced by carrageenan and lung fibrosis induced by bleomycin in mice.
  • Blocks metastasis in an experimental animal model of breast cancer metastasis.
  • Significantly decreases the numbers of hepatic micrometastases in vivo.