Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Msx-127 (NSC-23026) | 6616-56-4 | MFCD00087356 | 99.7% | 308.37 g/mol | C16H24N2O4 | 25 MG
SDP

Catalog No. 5000246312
Encompass_Preferred

MSX-127 is a small-molecule C-X-C chemokine receptor 4 (CXCR4) antagonist supplied for research use. It inhibits CXCR4-mediated signaling and has been used in studies of cancer metastasis. The compound is provided as a high-purity solid with documented storage and solubility properties suitable for in vitro research applications.

  • Acts as a CXCR4 antagonist relevant to cancer and immunology research.
  • High reported purity (≈99.68%).
  • Molecular weight 308.37 g/mol and formula C16H24N2O4.
  • White to off-white solid appearance.
  • Soluble in DMSO (≈41.67 mg/mL); ultrasonic assistance may be required.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).

Catalog No. 50-002-46312 Supplier Medchemexpress LLC Supplier No. HY10300925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MSX-127 is a small-molecule C-X-C chemokine receptor 4 (CXCR4) antagonist supplied for research use. It inhibits CXCR4-mediated signaling and has been used in studies of cancer metastasis. The compound is provided as a high-purity solid with documented storage and solubility properties suitable for in vitro research applications.

  • Acts as a CXCR4 antagonist relevant to cancer and immunology research.
  • High reported purity (≈99.68%).
  • Molecular weight 308.37 g/mol and formula C16H24N2O4.
  • White to off-white solid appearance.
  • Soluble in DMSO (≈41.67 mg/mL); ultrasonic assistance may be required.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.