Learn More
Enzo Life Sciences Msx 2 monoclonal antibody (2E12) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM010E
Encompass_Preferred
Clone: 2E12. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human Msx2 (N-terminal fragment). UniProt ID: P35548. Species reactivity: Human. Applications: IHC, WB. Recommended Dilutions: Immunohistochemistry (10µg/ml) Western Blot (1µg/ml) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7131