Learn More
Aviva Systems Biology Corp MSX1 Antibody - N-terminal region (ARP37094_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37094_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MSX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse MSX1. Alternate names: msh, Hox7, Hox-7, Hox7.1, AA675338, AI324650ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-56322