Learn More
Aviva Systems Biology Corp MSX1 Antibody - middle region (ARP31396_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31396_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MSX1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human MSX1. Alternate names: HOX7, HYD1, STHAG1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54205