Learn More
Aviva Systems Biology Corp MSX2 Antibody - N-terminal region (ARP38153_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38153_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MSX2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MSX2. Alternate names: FPP, MSH, PFM, CRS2, HOX8, PFM1ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56540