Learn More
Aviva Systems Biology Corp MSX2 Antibody - N-terminal region (ARP31398_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31398_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MSX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human MSX2. Alternate names: CRS2, FPP, HOX8, MSH, PFM, PFM1ApplicationIF, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54206