Learn More
New England Biolabs, Inc. Msz Exonuclease I - 1000 units
SDP
Catalog No. 502269742
Encompass
A DNA specific exonuclease isolated from Methylocaldum szegediense that catalyzes the removal of nucleotides from linear single-stranded DNA in the 3 prime to 5 prime direction with optimal activity between 45 and 60 degrees C. Isolated from a strain of E. coli which carries the cloned pMC45-1 gene from Methylocaldum szegediense.
This item is not returnable. View return policy
Catalog No. 50-226-9742 Supplier New England Biolabs, Inc. Supplier No. M0527S
A DNA specific exonuclease isolated from Methylocaldum szegediense that catalyzes the removal of nucleotides from linear single-stranded DNA in the 3 prime to 5 prime direction with optimal activity between 45 and 60 degrees C. Isolated from a strain of E. coli which carries the cloned pMC45-1 gene from Methylocaldum szegediense.