Learn More
Dojindo Molecular Technologies Inc MT-1 MitoMP Detection Kit allows monitoring of mitochondrial membrane potential with highly sensitive and applicable to live and fixation after staining (Detection Ex: 530-560 nm/Em: 570-640 nm) 20 µL x 3
SDP
Supplier:  Dojindo Molecular Technologies Inc MT1310
Encompass_Preferred
JC-1 dye, TMRE, and TMRM are widely used to monitor mitochondrial membrane potential. However, these dyes have some limitations such as low photostability and poor retention after aldehyde fixation. These limitations result in poor reproducibility of experiments. Since mitochondrial membrane potential fluctuates according to slight changes in cellular status, the use of fixed cells is important to obtain repeatable data for the measurement of mitochondrial membrane potential. MT-1 stained in live cells remains unquenched even if fixed with paraformaldehyde. Additionally, the Imaging Buffer included in this kit minimizes background fluorescence and maintains cell vitality while the assay is being performed. Dojindo offers various reagents for mitochondria research. (https://www.dojindo.com/mitochondria-research/)
Catalog No. NC1933275