Learn More
Cayman Chemical MT-1207 (hydrochloride), Purity: ≥98%, CAS: 1610794-70-1, 50 mg
SDP
Catalog No. 502943620
Encompass_Preferred
An α1-adrenergic and 5-HT2A receptor antagonist (IC50s = 2A- and α2B-adrenergic and 5-HT1A, 5-HT1B, 5-HT2B, and 5-HT2C receptors (IC50s = 18.1, 4.89, 4.68, 41, 11.5, and 26 nM, respectively), among others; decreases systolic and diastolic blood pressure and heart rate in spontaneously hypertensive rats and 2K-1C renal hypertensive dogs at 10 and 4 mg/kg, respectively
Catalog No. 50-294-3620 Supplier Cayman Chemical Supplier No. 4172050 MG
An α1-adrenergic and 5-HT2A receptor antagonist (IC50s = 2A- and α2B-adrenergic and 5-HT1A, 5-HT1B, 5-HT2B, and 5-HT2C receptors (IC50s = 18.1, 4.89, 4.68, 41, 11.5, and 26 nM, respectively), among others; decreases systolic and diastolic blood pressure and heart rate in spontaneously hypertensive rats and 2K-1C renal hypertensive dogs at 10 and 4 mg/kg, respectively