Learn More
Cayman Chemical MT-45-d11hydrochlorIdeex 100ug
SDP
Catalog No. 502915044
Encompass_Preferred
An internal standard for the quantification of MT-45 by GC- or LC-MS; regulated as a Schedule I compound in the United States; intended for research and forensic applications
Catalog No. 50-291-5044 Supplier Cayman Chemical Supplier No. 36876100 UG
An internal standard for the quantification of MT-45 by GC- or LC-MS; regulated as a Schedule I compound in the United States; intended for research and forensic applications