Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mt 63-78 | 1179347-65-9 | 98.0% | 326.35 g/mol | C21H14N2O2 | 10 MG
SDP

Catalog No. 5000210868
Encompass_Preferred

MT 63-78 is a small-molecule research compound that functions as a direct activator of AMP-activated protein kinase (AMPK) and inhibits mTORC1 signaling. It induces mitotic arrest and apoptosis, and has reported antitumor effects, including blocking prostate cancer growth via suppression of lipogenesis and mTORC1 pathways.

  • Direct AMPK activator with reported EC50 25 μM
  • Induces mitotic arrest and apoptosis in cellular assays
  • Reported antitumor activity, including inhibition of prostate cancer growth
  • Molecular weight 326.35 g/mol; formula C21H14N2O2
  • Purity 98.0%
  • Available in 1 mg, 5 mg, 10 mg, 50 mg, 100 mg packs; store protected from light at 4°C (in solvent: -80°C for 6 months, -20°C for 1 month)

Catalog No. 50-002-10868 Supplier Medchemexpress LLC Supplier No. HYW05884910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MT 63-78 is a small-molecule research compound that functions as a direct activator of AMP-activated protein kinase (AMPK) and inhibits mTORC1 signaling. It induces mitotic arrest and apoptosis, and has reported antitumor effects, including blocking prostate cancer growth via suppression of lipogenesis and mTORC1 pathways.

  • Direct AMPK activator with reported EC50 25 μM
  • Induces mitotic arrest and apoptosis in cellular assays
  • Reported antitumor activity, including inhibition of prostate cancer growth
  • Molecular weight 326.35 g/mol; formula C21H14N2O2
  • Purity 98.0%
  • Available in 1 mg, 5 mg, 10 mg, 50 mg, 100 mg packs; store protected from light at 4°C (in solvent: -80°C for 6 months, -20°C for 1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.