Learn More
Medchemexpress LLC MT-7716 hydrochloride | 1215859-93-0 | 99.1% | 477.0 | C27H29ClN4O2 | 1 ML
SDP
Catalog No. 5000239984
Encompass_Preferred
MT-7716 hydrochloride is the hydrochloride salt of a selective non-peptide nociceptin (NOP) receptor agonist used for preclinical pharmacology research. It is supplied at high purity and is offered both as a ready-to-use DMSO solution and as solid quantities for formulation and biological testing. The compound is commonly used in studies of alcohol dependence, withdrawal, and relapse mechanisms.
Catalog No. 50-002-39984 Supplier Medchemexpress LLC Supplier No. HY10709410MM 1ML
MT-7716 hydrochloride is the hydrochloride salt of a selective non-peptide nociceptin (NOP) receptor agonist used for preclinical pharmacology research. It is supplied at high purity and is offered both as a ready-to-use DMSO solution and as solid quantities for formulation and biological testing. The compound is commonly used in studies of alcohol dependence, withdrawal, and relapse mechanisms.