Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MT-7716 hydrochloride | 1215859-93-0 | 99.1% | 477.00 g/mol | C27H29ClN4O2 | 5 MG
SDP

Catalog No. 5000240374
Encompass_Preferred

MT-7716 hydrochloride is the hydrochloride salt of a selective, non-peptide nociceptin (NOP) receptor agonist used in preclinical pharmacology research. It is provided as a high-purity solid for use in in vitro assays and in vivo studies investigating alcohol abuse and relapse prevention.

  • High purity solid suitable for research applications.
  • Soluble in DMSO at ≥ 100 mg/mL for in vitro use.
  • Manufacturer-recommended in vivo formulations available for dosing.
  • Storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Molecular weight 477.00 g/mol; molecular formula C27H29ClN4O2.

Catalog No. 50-002-40374 Supplier Medchemexpress LLC Supplier No. HY1070945MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MT-7716 hydrochloride is the hydrochloride salt of a selective, non-peptide nociceptin (NOP) receptor agonist used in preclinical pharmacology research. It is provided as a high-purity solid for use in in vitro assays and in vivo studies investigating alcohol abuse and relapse prevention.

  • High purity solid suitable for research applications.
  • Soluble in DMSO at ≥ 100 mg/mL for in vitro use.
  • Manufacturer-recommended in vivo formulations available for dosing.
  • Storage: 4°C sealed; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Molecular weight 477.00 g/mol; molecular formula C27H29ClN4O2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.