Learn More
Medchemexpress LLC MT-7716 hydrochloride | 1215859-93-0 | MFCD07778657 | 99.1% | 477.00 | C27H29ClN4O2 | 25 MG
SDP
Catalog No. 5000239901
Encompass_Preferred
MT-7716 hydrochloride is the hydrochloride salt of MT-7716 (synonym W-212393), a selective non-peptide nociceptin receptor (NOP) agonist investigated as a potential treatment for alcohol abuse and relapse prevention. The compound is supplied as a solid with reported purity 99.06%, molecular formula C27H29ClN4O2, and molecular weight 477.00. It is offered in milligram quantities and as ready-to-use DMSO solutions, and should be stored sealed at 4°C (solutions at -80°C or -20°C per recommended timelines).
  • Selective nociceptin receptor (NOP) agonist for receptor pharmacology studies.
  • High reported purity (99.06%).
  • Available as a solid and as DMSO solutions for ready use.
  • Appropriate for preclinical pharmacology and relapse prevention research.
  • Store sealed and protected from moisture to maintain stability.
Catalog No. 50-002-39901 Supplier Medchemexpress LLC Supplier No. HY10709425MG
MT-7716 hydrochloride is the hydrochloride salt of MT-7716 (synonym W-212393), a selective non-peptide nociceptin receptor (NOP) agonist investigated as a potential treatment for alcohol abuse and relapse prevention. The compound is supplied as a solid with reported purity 99.06%, molecular formula C27H29ClN4O2, and molecular weight 477.00. It is offered in milligram quantities and as ready-to-use DMSO solutions, and should be stored sealed at 4°C (solutions at -80°C or -20°C per recommended timelines).
  • Selective nociceptin receptor (NOP) agonist for receptor pharmacology studies.
  • High reported purity (99.06%).
  • Available as a solid and as DMSO solutions for ready use.
  • Appropriate for preclinical pharmacology and relapse prevention research.
  • Store sealed and protected from moisture to maintain stability.