Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mt-7716 hydrochloride | 1215859-93-0 | MFCD07778657 | 99.1% | 477.00 g·mol⁻¹ | C27H29ClN4O2 | 100 MG
SDP

Catalog No. 5000240590
Encompass_Preferred

MT-7716 hydrochloride is a selective non-peptide nociceptin receptor (NOP) agonist supplied as the hydrochloride salt for research use. It is investigated for reducing alcohol-seeking behavior and relapse, and is provided as a solid reagent for in vitro and in vivo pharmacology studies.

  • Selective non-peptide nociceptin (NOP) receptor agonist.
  • Investigated for reduction of alcohol seeking and relapse prevention.
  • Provided as the hydrochloride salt in solid form for research applications.
  • High purity (≈99.1%) suitable for analytical and pharmacology studies.
  • Molecular formula C27H29ClN4O2, molecular weight 477.00 g·mol⁻¹.
  • Storage: 4°C sealed, away from moisture; in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-40590 Supplier Medchemexpress LLC Supplier No. HY107094100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

MT-7716 hydrochloride is a selective non-peptide nociceptin receptor (NOP) agonist supplied as the hydrochloride salt for research use. It is investigated for reducing alcohol-seeking behavior and relapse, and is provided as a solid reagent for in vitro and in vivo pharmacology studies.

  • Selective non-peptide nociceptin (NOP) receptor agonist.
  • Investigated for reduction of alcohol seeking and relapse prevention.
  • Provided as the hydrochloride salt in solid form for research applications.
  • High purity (≈99.1%) suitable for analytical and pharmacology studies.
  • Molecular formula C27H29ClN4O2, molecular weight 477.00 g·mol⁻¹.
  • Storage: 4°C sealed, away from moisture; in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.