Learn More
Hach Company Mt. Hardware Sp. Cap-2in. Sani
Catalog No. 5000207277 Shop All Hach Company Products
Encompass
Mounting Hardware Special Cap-2-inch SanitarySpare special cap for inductive conductivity sensor and 3700-series sensors For use with sanitary sensor and MH018S8SZ hardware
Catalog No. 50-002-07277 Supplier Hach Company Supplier No. 70F1037003
Mounting Hardware Special Cap-2-inch SanitarySpare special cap for inductive conductivity sensor and 3700-series sensors For use with sanitary sensor and MH018S8SZ hardware