Learn More
Aviva Systems Biology Corp MTA1 Antibody - C-terminal region (ARP37738_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37738_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MTA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human MTA1. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56415