Learn More
Aviva Systems Biology Corp MTA2 Antibody - C-terminal region (ARP43599_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43599_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MTA2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human MTA2. Alternate names: DKFZp686F2281, MTA1L1, PIDApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58582