Learn More
Aviva Systems Biology Corp Mta2 Antibody - N-terminal region (ARP37167_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37167_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against Mta2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AW550797, Mata1l1, Mta1l1, mmta2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-56331