Learn More
Aviva Systems Biology Corp MTA2 Antibody - middle region (ARP35743_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35743_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MTA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MTA2. Alternate names: DKFZp686F2281, MTA1L1, PIDApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55836