Learn More
Proteintech Group Inc MTAP Polyclonal antibody
SDP
Supplier:  Proteintech Group Inc 114751AP150UL
Encompass_Preferred
The MTAP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MTAP. This antibody recognizes human, rat, mouse antigen. The MTAP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Catalog No. 50-225-6368