Learn More
ABclonal Technology MTAP Rabbit mAb
SDP
Catalog No. 5000008148
Encompass_Preferred
This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage pathway of both adenine and methionine The encoded enzyme is deficient in many cancers Multiple alternatively spliced transcript variants have been described for this gene
Catalog No. 50-000-08148 Supplier ABclonal Technology Supplier No. A2090720UL
This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage pathway of both adenine and methionine The encoded enzyme is deficient in many cancers Multiple alternatively spliced transcript variants have been described for this gene