Learn More
BIO SB INC MTAP (RBT-MTAP), RMab, 0.5ml Concentrate
Supplier:  BIO SB INC BSB373905
Encompass
MTAP antibody is a rabbit monoclonal antibody derived from cell culture supernatant that is concentrated, dialyzed, filter sterilized and diluted in buffer pH 7.5, containing BSA and sodium azide as a preservative.
Catalog No. NC2855844