Learn More
Medchemexpress LLC Mtb ATP synthase-IN-1 | 2642394-38-3 | 98.6% | 323.30 | 100 MG
SDP
Catalog No. 5000363103
Encompass_Preferred
Mtb ATP synthase-IN-1 (compound 6ab) is identified as a potent Mycobacterium tuberculosis (Mtb) ATP synthase inhibitor, demonstrating a MIC of 0.452-0.499 μg/mL against Mtb. This compound exhibits favorable metabolic stability, low cytotoxicity (with a Vero IC50 > 64 μg/mL), and acceptable oral bioavailability. It is suitable for research applications focused on anti-mycobacterium studies.
  • Potent Mycobacterium tuberculosis (Mtb) ATP synthase inhibitor
  • MIC of 0.452-0.499 μg/mL against Mtb
  • Good metabolic stability
  • Low cytotoxicity (Vero IC50 > 64 μg/mL)
  • Acceptable oral bioavailability
  • Intended for researching anti-mycobacterium
Catalog No. 50-003-63103 Supplier Medchemexpress LLC Supplier No. HY146388100MG
Mtb ATP synthase-IN-1 (compound 6ab) is identified as a potent Mycobacterium tuberculosis (Mtb) ATP synthase inhibitor, demonstrating a MIC of 0.452-0.499 μg/mL against Mtb. This compound exhibits favorable metabolic stability, low cytotoxicity (with a Vero IC50 > 64 μg/mL), and acceptable oral bioavailability. It is suitable for research applications focused on anti-mycobacterium studies.
  • Potent Mycobacterium tuberculosis (Mtb) ATP synthase inhibitor
  • MIC of 0.452-0.499 μg/mL against Mtb
  • Good metabolic stability
  • Low cytotoxicity (Vero IC50 > 64 μg/mL)
  • Acceptable oral bioavailability
  • Intended for researching anti-mycobacterium