Learn More
Medchemexpress LLC Mtb ATP synthase-IN-1 | 2642394-38-3 | 98.6% | 323.30 | 50 MG
SDP
Catalog No. 5000363836
Encompass_Preferred
Mtb ATP synthase-IN-1 (compound 6ab) is a potent Mycobacterium tuberculosis (Mtb) ATP synthase inhibitor, with a MIC of 0.452-0.499 μg/mL against Mtb. It demonstrates good metabolic stability, low cytotoxicity (Vero IC50 > 64 μg/mL), and acceptable oral bioavailability. This compound is suitable for researching anti-mycobacterium applications.
  • Potent Mtb ATP synthase inhibitor
  • MIC of 0.452-0.499 μg/mL against Mtb
  • Good metabolic stability
  • Low cytotoxicity (Vero IC50 > 64 μg/mL)
  • Acceptable oral bioavailability
  • Suitable for anti-mycobacterium research
Catalog No. 50-003-63836 Supplier Medchemexpress LLC Supplier No. HY14638850MG
Mtb ATP synthase-IN-1 (compound 6ab) is a potent Mycobacterium tuberculosis (Mtb) ATP synthase inhibitor, with a MIC of 0.452-0.499 μg/mL against Mtb. It demonstrates good metabolic stability, low cytotoxicity (Vero IC50 > 64 μg/mL), and acceptable oral bioavailability. This compound is suitable for researching anti-mycobacterium applications.
  • Potent Mtb ATP synthase inhibitor
  • MIC of 0.452-0.499 μg/mL against Mtb
  • Good metabolic stability
  • Low cytotoxicity (Vero IC50 > 64 μg/mL)
  • Acceptable oral bioavailability
  • Suitable for anti-mycobacterium research