Learn More
Medchemexpress LLC Mtb ATP synthase-IN-1 | 2642394-38-3 | 98.6% | C17H13N3O4 | 10MG
SDP
Catalog No. 5000258172
Encompass_Preferred
Mtb ATP synthase-IN-1 is a potent Mycobacterium tuberculosis ATP synthase inhibitor (compound 6ab) for anti-mycobacterial research. It exhibits MIC 0.452-0.499 μg/mL against M. tuberculosis, demonstrates good metabolic stability and acceptable oral bioavailability, and shows low cytotoxicity in Vero cells.
  • Potent inhibition of M. tuberculosis ATP synthase
  • MIC 0.452-0.499 μg/mL against M. tuberculosis
  • Good metabolic stability
  • Low cytotoxicity (Vero IC50 > 64 μg/mL)
  • High purity (~98.6%)
  • Available in multiple solid and solution pack sizes
  • Stable storage recommendations for powder and solutions
Catalog No. 50-002-58172 Supplier Medchemexpress LLC Supplier No. HY14638810MG
Mtb ATP synthase-IN-1 is a potent Mycobacterium tuberculosis ATP synthase inhibitor (compound 6ab) for anti-mycobacterial research. It exhibits MIC 0.452-0.499 μg/mL against M. tuberculosis, demonstrates good metabolic stability and acceptable oral bioavailability, and shows low cytotoxicity in Vero cells.
  • Potent inhibition of M. tuberculosis ATP synthase
  • MIC 0.452-0.499 μg/mL against M. tuberculosis
  • Good metabolic stability
  • Low cytotoxicity (Vero IC50 > 64 μg/mL)
  • High purity (~98.6%)
  • Available in multiple solid and solution pack sizes
  • Stable storage recommendations for powder and solutions