Learn More
Regis Technologies Inc GC Regeant, Regis, MTBSTFA + 1% TBDMCS (10 x 1 GM)
SDP
Catalog No. 50442893
Encompass
MtBSTFA is a Silylation Reagent. MtBSTFA replaces active hydrogens to form t-BDMS derivatives. Derivatization is usually complete upon dissolution with this exceptionally strong, yet mild silylating reagent. MtBSTFA derivatives are 104 times more stable to hydrolysis than their corresponding TMS derivatives and produce easily interpreted mass spectra for GC/MS. Addition of t-BDMCS catalyzes reactions of hindered alcohols and amines.
Catalog No. 50-442-893 Supplier Regis Technologies Inc Supplier No. 1270144200
MtBSTFA is a Silylation Reagent. MtBSTFA replaces active hydrogens to form t-BDMS derivatives. Derivatization is usually complete upon dissolution with this exceptionally strong, yet mild silylating reagent. MtBSTFA derivatives are 104 times more stable to hydrolysis than their corresponding TMS derivatives and produce easily interpreted mass spectra for GC/MS. Addition of t-BDMCS catalyzes reactions of hindered alcohols and amines.