Learn More
MSC MTC Bio Transfer Pipette, 4.5 mL, Graduated to 2 mL, Sterile, Individually Wrapped
Catalog No. 5000534137
Encompass
These unbreakable, one-piece transfer pipettes, made from ultra-clear, low-density polyethylene (LDPE), prevent contamination from rubber bulbs, broken glass, and infectious materials. Ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Sterile versions are excellent for tissue culture, with a low-affinity surface to minimize sample, cell, or protein loss.
  • Constructed from ultra-clear, low-density polyethylene (LDPE).
  • One-piece, unbreakable design.
  • Heat-sealable for airtight sample storage.
  • Provides uniform drop size for reproducible spotting and dispensing.
  • Ideal for microbiology, blood banking, hematology, immunology, and urinalysis.
  • Minimizes exposure to infectious materials.
  • Low-affinity surface reduces sample, cell, or protein loss.
  • RNase, DNase, and pyrogen-free.
Catalog No. 50-005-34137 Supplier MSC Supplier No. 00450114
These unbreakable, one-piece transfer pipettes, made from ultra-clear, low-density polyethylene (LDPE), prevent contamination from rubber bulbs, broken glass, and infectious materials. Ideal for microbiology, blood banking, hematology, immunology, and urinalysis. Sterile versions are excellent for tissue culture, with a low-affinity surface to minimize sample, cell, or protein loss.
  • Constructed from ultra-clear, low-density polyethylene (LDPE).
  • One-piece, unbreakable design.
  • Heat-sealable for airtight sample storage.
  • Provides uniform drop size for reproducible spotting and dispensing.
  • Ideal for microbiology, blood banking, hematology, immunology, and urinalysis.
  • Minimizes exposure to infectious materials.
  • Low-affinity surface reduces sample, cell, or protein loss.
  • RNase, DNase, and pyrogen-free.