Learn More
Hach Company Intellical™ MTC306 Combined Silver Metal Electrode for AT1000 Argentometry Titrations
Catalog No. 501998061 Shop All Hach Company Products
Encompass
Intellical™ MTC306 is a digital, combination silver metal rod probe with refillable reference and build-in temperature sensor. The probe is available with a 1 meter cable and is intended for titration use with Titralab AT1000 Series. The MTC306 is ideal for Argentometry precipitation titration determining chlorides/salt content in water and Food products applications.
Catalog No. 50-199-8061 Supplier Hach Company Supplier No. MTC30601
Intellical™ MTC306 is a digital, combination silver metal rod probe with refillable reference and build-in temperature sensor. The probe is available with a 1 meter cable and is intended for titration use with Titralab AT1000 Series. The MTC306 is ideal for Argentometry precipitation titration determining chlorides/salt content in water and Food products applications.