Learn More
Aviva Systems Biology Corp Mtch1 Antibody - middle region (ARP47012_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47012_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mtch1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Mtch1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-279