Learn More
Cayman Chemical MTDB-Alkyne, Purity: ≥98%, CAS: 2923694-73-7, 1 mg
SDP
Catalog No. 502918461
Encompass_Preferred
A clickable RNA pseudoknot binder; has been used in the synthesis of PINAD molecules for the degradation of SARS-CoV-2 RNA
Catalog No. 50-291-8461 Supplier Cayman Chemical Supplier No. 389261 MG
A clickable RNA pseudoknot binder; has been used in the synthesis of PINAD molecules for the degradation of SARS-CoV-2 RNA