Learn More
Enzo Life Sciences MTEP (25mg). CAS: 329205-68-7
SDP
Supplier:  Enzo Life Sciences ALX550381M025
Encompass_Preferred
Potent and highly selective antagonist for mGluR5 with a 5-fold higher anxiolytic activity than MPEP (ALX-550-368). Shows fewer off-target effects than MPEP. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO, 100% ethanol or methanol. Long Term Storage: +4°C.
Catalog No. 50-200-9550